Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MATH2/NEUROD6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP229865 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229865 0.1 mg
NB019492 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229865 Supplier Novus Biologicals Supplier No. NBP229865
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MATH2/NEUROD6 Polyclonal specifically detects MATH2/NEUROD6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MATH2/NEUROD6
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200-1:1000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin
Formulation 50% saturated ammonium sulfate and PBS with No Preservative
Gene Accession No. Q96NK8
Gene Alias Atoh2, BHLHA2, bHLHa2MATH2, Class A basic helix-loop-helix protein 2, Math-2, NeuroD6, neurogenic differentiation 6, neurogenic differentiation factor 6, NEX1M, Protein atonal homolog 2
Gene Symbols NEUROD6
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63974
Test Specificity Recognizes NeuroD 6 (MATH2, NEX1), a Helix-loop-helix class of transcription factor.
Target Species Human, Mouse
Content And Storage Store at -70°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.