Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MATN3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP304918100UL

 View more versions of this product

Catalog No. NB096183


Only null left
Add to Cart

Description

Description

MATN3 Polyclonal specifically detects MATN3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MATN3
Polyclonal
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
4148
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
DIPOA, EDM5, HOA, matrilin 3, matrilin-3, OADIP, OS2
Recombinant fusion protein containing a sequence corresponding to amino acids 277-486 of human MATN3 (NP_002372.1).
100 μL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.