Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MATN3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
| Antigen | MATN3 |
|---|---|
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
MATN3 Polyclonal specifically detects MATN3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| MATN3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 4148 | |
| IgG | |
| Affinity purified |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DIPOA, EDM5, HOA, matrilin 3, matrilin-3, OADIP, OS2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 277-486 of human MATN3 (NP_002372.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title