Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrilin-1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176325 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176325 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176325 Supplier Novus Biologicals Supplier No. NBP176325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Matrilin-1 Polyclonal specifically detects Matrilin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Matrilin-1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_002370
Gene Alias cartilage matrix protein, matrilin 1, cartilage matrix protein
Gene Symbols MATN1
Host Species Rabbit
Immunogen Antibody was raised against a 12 amino acid synthetic peptide from near the amino terminus of human MATN1. The immunogen is located within amino acids 120 - 170 of MATN1. Amino Acid Squence: EGGRSRSPDISK
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4146
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.