Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrin 3 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001761 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001761 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001761 Supplier Novus Biologicals Supplier No. NB1001761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 11 publications

Matrin 3 Polyclonal specifically detects Matrin 3 in Human, Mouse samples. It is validated for Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen Matrin 3
Applications Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:15000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:250-1:1000, Immunoprecipitation 1-4 μg/mg of lysate, Immunohistochemistry-Paraffin 1:10-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. P43243
Gene Alias distal 2, DKFZp686K0542, DKFZp686K23100, matrin 3, matrin-3, vocal cord and pharyngeal weakness with distal myopathy
Gene Symbols MATR3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 800 and the C-terminus (residue 847) of human Matrin 3 using the numbering given in entry NP_061322.2 (GeneID 9782).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9782
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, Small-eared galago, Little brown bat and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.