Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matriptase/ST14 Antibody (416802), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB371169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371169 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371169 Supplier Novus Biologicals Supplier No. FAB3946I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Matriptase/ST14 Monoclonal specifically detects Matriptase/ST14 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen Matriptase/ST14
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 416802
Conjugate Janelia Fluor 549
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, HAI, Matriptase, Membrane-type serine protease 1, MT-SP1EC 3.4.21.109, prostamin, PRSS14, Serine protease 14, Serine protease TADG-15, SNC19MTSP1, suppression of tumorigenicity 14 (colon carcinoma), suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin), suppressor of tumorigenicity 14 protein, TADG15, TMPRSS14, tumor associated differentially expressed gene 15 protein, Tumor-associated differentially-expressed gene 15 protein
Host Species Mouse
Immunogen E. coli-derived recombinant human Matriptase/ST14 Catalytic Domain, Val615-Val855, Accession # Q9Y5Y6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6768
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.