Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matriptase/ST14 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB292425 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292425 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292425 Supplier Novus Biologicals Supplier No. NB1002308PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Matriptase/ST14 Polyclonal antibody specifically detects Matriptase/ST14 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Matriptase/ST14
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias EC 3.4.21, HAI, Matriptase, Membrane-type serine protease 1, MT-SP1EC 3.4.21.109, prostamin, PRSS14, Serine protease 14, Serine protease TADG-15, SNC19MTSP1, suppression of tumorigenicity 14 (colon carcinoma), suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin), suppressor of tumorigenicity 14 protein, TADG15, TMPRSS14, tumor associated differentially expressed gene 15 protein, Tumor-associated differentially-expressed gene 15 protein
Host Species Rabbit
Immunogen Maps to a region between residues 800 and the C-terminus (residue 855) of human Membrane-type serine protease 1 using the numbering given in SwissProt entry Q9Y5Y6 (GeneID 6768).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6768
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.