Learn More
Thermo Scientific™ Matrix™ 1.0mL ScrewTop Tubes in Barcoded Latch Racks
GREENER_CHOICE
Catalog No. 14754584
Change view
Click to view available options
Product Type:
Screw Top Tube
Screw Top with White Patch
Color:
Amber
Clear
Closure Color:
Blue
Colorless
Green
Not Included
Purple
Red
White
Yellow
Fit for Purpose:
Sterile and PCR Ready
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color Closure Color Fit for Purpose
14-754-584 Screw Top with White Patch Clear Not Included Sterile and PCR Ready
14-754-563 Screw Top Tube Clear Colorless Sterile and PCR Ready
14-754-571 Screw Top Tube Clear White Sterile and PCR Ready
14-754-562 Screw Top Tube Clear Blue Sterile and PCR Ready
14-754-581 Screw Top Tube Clear Not Included Sterile and PCR Ready
14-754-567 Screw Top Tube Clear Purple Sterile and PCR Ready
14-754-565 Screw Top Tube Clear Green Sterile and PCR Ready
14-754-560 Screw Top Tube Amber Red Sterile and PCR Ready
14-754-569 Screw Top Tube Clear Red Sterile and PCR Ready
14-754-580 Screw Top Tube Amber Not Included Sterile and PCR Ready
14-754-577 Screw Top Tube Clear Yellow Sterile and PCR Ready
14-754-575 Screw Top with White Patch Clear Colorless Sterile and PCR Ready
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 14-754-584 Supplier Thermo Scientific™ Supplier No. 3742WPBR
Only null left
Add to Cart
Add to Cart
Track your samples easily at low temperatures, including vapor phase Liquid Nitrogen with 1.0mL ScrewTop Tubes in barcoded latch racks.
Securely store samples at low temperatures, including vapor phase Liquid Nitrogen with these 1.0mL ScrewTop Tubes in barcoded latch racks. Tubes are molded from medical grade resins and feature a permanent, high contrast 2D barcode. These tubes are packed in our Barcoded Latch Racks that feature a linear barcode, 2D barcode and human readable rack identification on three sides of the rack. Use in combination with Thermo Scientific barcode readers, arrayers and capping equipment. The 2D screwtop tubes meet a range of manual and automated long term storage needs; barcoded racks make it easy to track the rack as well.
Dependable Sample Protection
  • Tubes are constructed of virgin class VI medical grade polypropylene
  • ScrewTop caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy ABS to protect tubes in case of drops
  • ScrewTop tubes are compatible with storage down to -196°C
Secure Tracking
  • Permanent, unique 2D tube barcode is laser-etched on bottom of each tube
  • Racks feature linear barcode, human readable ID and 2D barcode for ultimate sample tracking
  • 2D barcode on tubes allow you to scan and decode tubes all at once without removing them from the rack, and allows data to be associated with each tube
  • Tube and rack are compatible with Thermo Scientific™ VisionMate™ readers
Superior Storage Format
  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Several capping / decapping options available to manipulate a single tube, 8 tubes, or 96 tubes at once
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Product Type Screw Top with White Patch
Closure Color Not Included
Autoclavable Not Autoclavable
Capacity (Metric) 1.0 mL
Bottom Shape V-bottom
Material Polypropylene (Medical Grade)
Packaging 5 Latch Racks of 96 Tubes
Barcode Tube barcode: Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction; Rack barcode: 2D Datamatrix; Linear EAN Code 128; Human Readable
Product Line Matrix
Sterility Sterile
Closure Included No
Capacity (English) 0.0338 fl oz.
Fit for Purpose Sterile and PCR Ready