Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAVS Antibody (Adri-1), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB172602 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172602 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172602 Supplier Novus Biologicals Supplier No. NBP280052V
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAVS Monoclonal antibody specifically detects MAVS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen MAVS
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone Adri-1
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051
Host Species Mouse
Immunogen Recombinant human MAVS (aa 160-450).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune Dysfunction, Immune System Diseases, Immunology, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 57506
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.