Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAVS Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB356121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356121 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356121 Supplier Novus Biologicals Supplier No. NBP177275MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAVS Polyclonal antibody specifically detects MAVS in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ChIP assay (ChIP)

Specifications

Antigen MAVS
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), ChIP Assay
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP)
Formulation 50mM Sodium Borate
Gene Alias CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide from near the amino terminus of human VISA. The immunogen is located within the first 50 amino acids of VISA. Amino Acid Squence: CLTARDQDRLRAT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune Dysfunction, Immune System Diseases, Immunology, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 57506
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.