Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAVS Antibody (OTI9C4), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N272593J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272593J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272593J646 Supplier Novus Biologicals Supplier No. NBP272593JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAVS Monoclonal specifically detects MAVS in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MAVS
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI9C4
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051
Gene Symbols MAVS
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-247 of human MAVS (NP_065797) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Immune Dysfunction, Immune System Diseases, Immunology, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 57506.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.