Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Max Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB322693 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322693 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322693 Supplier Novus Biologicals Supplier No. NBP300047PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Max Polyclonal antibody specifically detects Max in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Max
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation PBS
Gene Alias BHLHD4, bHLHd4MGC11225, bHLHd5, bHLHd6, bHLHd7, bHLHd8, Class D basic helix-loop-helix protein 4, helix-loop-helix zipper protein, MAX protein, MGC10775, MGC18164, MGC34679, MGC36767, MYC associated factor X, Myc-associated factor X, orf1, protein max
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Max (Accession#: NP_002373; Met1-Ser160)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4149
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.