Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAX binding protein Antibody, Novus Biologicals™
SDP

Catalog No. NBP204052 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP204052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP204052 Supplier Novus Biologicals Supplier No. NBP204052
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAX binding protein Polyclonal specifically detects MAX binding protein in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen MAX binding protein
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_064706.1
Gene Alias BHLHD3, bHLHd3Max-interacting protein, Class D basic helix-loop-helix protein 3, MAD6, MAX binding protein, MXD6, Myc antagonist MNT, Protein ROX, ROXmax-binding protein MNT
Gene Symbols MNT
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human MAX Binding Protein using the numbering given in entry NP_064706.1 (GeneID 4335).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4335
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.