Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maxi Potassium channel beta Antibody, Novus Biologicals™
SDP

Catalog No. NB300535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300535 Supplier Novus Biologicals Supplier No. NB300535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

Maxi Potassium channel beta Polyclonal specifically detects Maxi Potassium channel beta in Human, Mouse, Rat, Porcine, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen Maxi Potassium channel beta
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2.0 ug/ml, Flow Cytometry, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1:200 - 1:2000, Knockdown Validated
Formulation PBS, 1 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. Q16558
Gene Alias BK channel beta subunit, BK channel subunit beta-1, BKbeta, BKbeta1, Calcium-activated potassium channel subunit beta, calcium-activated potassium channel subunit beta-1, Calcium-activated potassium channel, subfamily M subunit beta-1, Charybdotoxin receptor subunit beta-1, Hbeta1, hslo-beta, K(VCA)beta, k(VCA)beta-1, large conductance Ca2+-activated K+ channel beta 1 subunit, Maxi K channel beta subunit, Maxi K channel subunit beta-1, potassium large conductance calcium-activated channel, subfamily M, beta member1, Slo-beta, Slo-beta-1
Gene Symbols KCNMB1
Host Species Rabbit
Immunogen Synthetic Peptide: L(90) Y H T E D T R D Q N Q Q C(103)
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Potassium Channels
Primary or Secondary Primary
Gene ID (Entrez) 3779
Target Species Human, Mouse, Rat, Pig, Canine, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.