Learn More
3M™ Maxim™ Elite 1000 Series Safety Glasses
3M™ Maxim™ Elite 1000 Series Safety Glasses with ANSI Z87.1-2015 safety compliance
$476.21 - $636.00
Specifications
Certifications/Compliance ANSI Z87.1-2015
For Use With (Application) Primary Metals, Chemicals, Automotive, Masonary, UV Protection, Grinding, Facility Maintenance, Construction, Sub Applications, Transportation, Assembly, Remediation, Machine Operations, Law Enforcement, Chemical Handling, Pharmaceuticals, Machining, Mining, Riveting, Food and Beverage, Drilling, Food and Beverage Manufacturing, Aviation, Pouring/Casting, Chiseling, Sawing, General Manufacturing, Power Fastening, Oil and Gas, Demolition
Lens Material Polycarbonate
Frame Type Full Frame
View More Specs
Products 7
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
19-072-202
View Documents
3M™
7100222267
Blue, Mirror
Case of 20 for $497.00
Only null left
Add to Cart
19-072-195
View Documents
3M™
7100222270
Purple, Mirror
Case of 20 for $519.05
Only null left
Add to Cart
19-072-198
View Documents
3M™
7100222271
Brown
Case of 5 for $636.00
Only null left
Add to Cart
19-072-203
View Documents
3M™
7100222265
Clear
Case of 20 for $532.61
Only null left
Add to Cart
19-072-201
View Documents
3M™
7100222268
Light Gray
Case of 20 for $476.21
Only null left
Add to Cart
19-072-200
View Documents
3M™
7100222266
Gray
Case of 20 for $532.61
Only null left
Add to Cart
19-072-196
View Documents
3M™
7100222264
Photochromic N/A N/A N/A
Description
Description
  • The next generation Maxim design offers upgraded features with perfomance lens technology
  • Influenced by lifestyle eyewear with stylish wrap-around lens design
  • Lightweight rubber painted matte frame material with embossed 3M logo
  • Blue or blue-violet mirrored lens options increase sunlight reflection, perfect for working in bright conditions
  • 3M eye protection meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
  • Polycarbonate lenses absorb 99.9% of UVA and UVB rays
  • Model with brown lens is polarized
Specifications
Specifications
ANSI Z87.1-2015
Primary Metals, Chemicals, Automotive, Masonary, UV Protection, Grinding, Facility Maintenance, Construction, Sub Applications, Transportation, Assembly, Remediation, Machine Operations, Law Enforcement, Chemical Handling, Pharmaceuticals, Machining, Mining, Riveting, Food and Beverage, Drilling, Food and Beverage Manufacturing, Aviation, Pouring/Casting, Chiseling, Sawing, General Manufacturing, Power Fastening, Oil and Gas, Demolition
Polycarbonate
Full Frame
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.