Learn More
Thermo Scientific™ Maxima H Minus First Strand cDNA Synthesis Kit PROMO
Maxima H Minus First Strand cDNA Synthesis Kit, available with or without dsDNase, is a complete system for highly efficient synthesis of first strand cDNA.
Specifications
Content And Storage | Maxima H Minus Enzyme Mix, Oligo(dT)18 and Random hexamer primers, 5X RT Buffer, dNTP Mix, Nuclease-free water, Store at -20°C. |
---|---|
Format | Kit |
GC-Rich PCR Performance | High |
Reaction Speed | 30 min. |
Technique | Reverse Transcription |
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
FERK1651PM
|
Thermo Scientific™
K1651 |
20 Reactions | N/A | N/A | N/A | ||||
FERK1652PM
|
Thermo Scientific™
K1652 |
100 Reactions | N/A | N/A | N/A | ||||
Description
This kit allows synthesis of long cDNAs up to 20 kb at elevated temperatures (up to 65°C), superseding other systems' abilities to produce full-length cDNA. Due to increased synthesis rates the reaction can be completed in 30 minutes.
For reverse transcription the kit uses Maxima H Minus Reverse Transcriptase (RT), which is an advanced enzyme derived by in vitroevolution of MMLV RT.
The Maxima H Minus First Strand cDNA Synthesis Kit with dsDNase provides a dramatically simplified workflow that combines genomic DNA elimination and cDNA synthesis into one-tube procedure. The kit contains a novel double-strand-specific DNase (dsDNase) engineered to remove contaminating genomic DNA from RNA preps in two minutes without damage to quality or quantity of RNA. Highly specific dsDNase activity towards double-stranded DNA ensures that single-stranded DNA (such as cDNA and primers) is not cleaved, and dsDNase treated RNA can be directly added to reverse transcription.
Features of the Maxima H Minus First Strand cDNA Synthesis Kit include:
- Increased reaction temperatures–the first strand of cDNA can be synthesized within the 42-65°C temperature range
- High yields of full-length first strand cDNA–with RNA templates up to 20 kb
- Flexible priming–oligo(dT)18, random hexamer or gene-specific primers
- Integrated genomic DNA removal step with dsDNase kit
Applications
- First Strand cDNA synthesis for RT-PCR
- Construction of cDNA libraries
- Generation of probes for hybridization
- Antisense RNA synthesis
Contents
- Maxima H Minus Enzyme Mix
- Oligo(dT)18 and Random hexamer primers
- 5X RT Buffer
- dNTP Mix
- Nuclease-free water
First Strand Synthesis Kit with dsDNase also contains
- dsDNase and 10X dsDNase Buffer
Additional information about reaction components
- Maxima H Minus Enzyme Mix contains Maxima H Minus Reverse Transcriptase and RiboLock RNase Inhibitor. RiboLock RNase Inhibitor effectively protects RNA templates from degradation by RNases A, B, and C at temperatures up to 55°C.
- Oligo(dT)18 and random hexamer primers are supplied with the kit. Random hexamer primers bind non-specifically and are used to synthesize cDNA from all RNAs in a total RNA population. The oligo(dT)18 primer selectively anneals to the 3'-end of poly(A) RNA, synthesizing cDNA only from poly(A) tailed mRNA. Gene-specific primers may also be used with the kit to prime synthesis from a specified sequence.
- 10 mM dNTP Mix is a premixed aqueous solution of dATP, dTTP, dCTP, and dGTP.
- Nuclease-free water is provided for reaction set-up and dilution of sample DNA. The absence of endo-, exodeoxyribonucleases, ribonucleases, and phosphatases has been confirmed by appropriate quality tests.
Specifications
Maxima H Minus Enzyme Mix, Oligo(dT)18 and Random hexamer primers, 5X RT Buffer, dNTP Mix, Nuclease-free water, Store at -20°C. | |
High | |
Reverse Transcription | |
Maxima H Minus | |
Dry Ice | |
First-Strand cDNA | |
Reverse Transcription | |
RNA |
Kit | |
30 min. | |
50°C to 55°C | |
Reduced | |
Real Time PCR (qPCR), RT-PCR | |
Separate components | |
Up to 20 kb | |
Kit only |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.