Learn More
Promega Maxwell™ 16 LEV Plant RNA Kit
Catalog No. PRAS1430 Shop All Promega Corporation Products
Click to view available options
Quantity:
48 Preps
The Maxwell 16 LEV Plant RNA Kit is used with the Maxwell 16 Research Instrument (Cat.# AS2000) to provide an easy method for efficient, automated purification of RNA from plant tissue samples.
  • Automated RNA Extraction from 1-16 Plant Tissue Samples in 60 Minutes
  • Extracts RNA from corn, soybean and Arabidopsis tissue samples
  • Purified RNA can be used directly in downstream applications, including RT-qPCR
  • Less variability than CTAB and manual-spin-column methods
Specifications
Content And Storage 15°C to 30°C
Quantity 48 Preps
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.