Learn More
Promega Maxwell™ 16 Purification Instruments
Shop All Promega Corporation ProductsDescription
Includes
Maxwell 16 instrument includes power cable and RS-232 cable for firmware upgrades. SEV Hardware Kit includes Maxwell 16 magnetic rod assembly, Maxwell 16 cartridge rack, Maxwell 16 magnetic elution rack. LEV Hardware Kit includes LEV magnetic rod assembly, LEV plunger bar adapter, Maxwell 16 LEV cartridge rack. Maxwell 16 MDx SEV includes a bar code reader, software to track and transfer data, and a UV bulb for decontamination.
Requires
Either the SEV kit (Mfr. No. AS1200) or the LEV kit (Mfr. No. AS1250) to complete a functional instrument. Also requires Maxwell 16 Purification Kits.
Purify samples using MagneSil™ Paramagnetic Particles, allowing optimal capture, washing, and elution of the target material. The magnetic-particle-handling instrument vigorously mixes samples with enough force to grind tissue in reagent cartridge without clogged tips and partial reagent transfers that can cause suboptimal results.
- Reduce hands-on time to purified sample and improve consistency of yield and purity
- Provide preprogrammed, sample-specific automated methods
- Process up to 16 samples in 30 to 45 minutes, depending on sample type
- Eliminate preprocessing of mouse tails or tissue samples for gDNA purification
- Flexi-method firmware allows users to define key instrument operating parameters to simultaneously purify up to 16 samples, without user intervention
- Use minimal bench space; footprint comparable to open laptop computer: 32.55 × 43.82 × 32.65cm
Choose Maxwell 16 SEV Hardware Kit or LEV Hardware Kit to convert the instrument to Standard or Low Elution mode, respectively
- Maxwell 16 SEV Hardware Kit is required to convert the Maxwell 16 LEV instrument to a 200 to 400μL standard elution volume (SEV) instrument for maximum yield
- Maxwell 16 LEV Hardware Kit is required to convert a Maxwell 16 SEV instrument to a 30 to 100μL low elution volume (LEV) instrument for maximum concentration
- Lets you optimize multiple instrument parameters to tailor instrument operation to your unique needs
- Allows modification of five key parameters: lysis time, binding, drying, elution, paramagnetic particle capture
- Includes a bar code reader, software to track and transfer data, and a UV bulb for decontamination
Certifications
All Maxwell 16 instruments are labeled GPLE (General Purpose Laboratory Equipment) in the U.S.
Specifications
Specifications
| For Use With (Application) | Automated purification of genomic DNA, RNA or recombinant Proteins from a wide range of sample types |
| Includes | Power cable, RS232 cable for firmware upgrades, CD with operating manual and warranty information, SEV Magnetic rod/plunger bar assembly, Maxwell™ 16 SEV cartridge rack, Maxwell™ 16 SEV Magnetic Elution rack, SEV and LE |
| Product Type | Maxwel 16 AS2000 instrument |
| Quantity | Each |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.