Learn More
Description
Includes
LEV Magnetic rod assembly, LEV plunger bar adaptor, Maxwell™ 16 LEV cartridge rack, quick start guide
Requires
Either the SEV kit (Mfr. No. AS1200) or the LEV kit (Mfr. No. AS1250) to complete a functional instrument. Also requires Maxwell 16 Purification Kits.
- Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
- Standard Elution Volume (SEV) is 200-400μL
- Low Elution Volume (LEV) is 30-100μL
- Plungers and Elution Tubes for the Maxwell(R) 16 Instruments only (AS2000 and AS3000)
Certifications
All Maxwell 16 instruments are labeled GPLE (General Purpose Laboratory Equipment) in the U.S.
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| For Use With (Equipment) | Maxwel 16 AS2000 instrument |
| For Use With (Application) | Automated purification of genomic DNA, RNA or recombinant Proteins from a wide range of sample types |
| Includes | Maxwell 16 instrument includes power cable and RS-232 cable for firmware upgrades. SEV Hardware Kit includes Maxwell 16 magnetic rod assembly, Maxwell 16 cartridge rack, Maxwell 16 magnetic elution rack. LEV Hardware Kit includes LEV magnetic rod assembl |
| Product Type | LEV Hardware Kit |
| Quantity | 1/Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.