Learn More
Promega Maxwell™ CSC RNA FFPE Kit
Shop All Promega Corporation Products
Click to view available options
Quantity:
48 Preps
Description
- Automated RNA Purification from FFPE Tissue Samples for Downstream In Vitro Diagnostic Assays
- Purifies RNA from formalin-fixed, paraffin-embedded (FFPE) colon, breast and lung tissues
- Purifies high-quality RNA that is suitable for use in amplification-based in vitro diagnostic assays
- Offers clinical customers a high-quality RNA purification method manufactured under cGMP
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Quantity | 48 Preps |