Learn More
Promega Maxwell™ RSC simplyRNA Cells Kit
Shop All Promega Corporation Products
Click to view available options
Quantity:
48 Preps
Description
- Automated RNA Purification from Cells using a Low Elution Volume
- Purifies total RNA from fresh or frozen cells
- Reduces pre-extraction sample handling to 4 steps
- No beta-mercaptoethanol needed
Specifications
Specifications
Content And Storage | 15°C to 30°C |
Quantity | 48 Preps |