Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maz Monoclonal Antibody (133), Active Motif™
SDP

Mouse Monoclonal Antibody

$156.65 - $638.65

Specifications

Antigen Maz
Clone 133
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2978
SDP
Encompass
View Documents
Active Motif
39936
10 μg
Each for $156.65
Only null left
Add to Cart
 
50-199-2977
SDP
Encompass
View Documents
Active Motif
39935
100 μg
Each for $638.65
Only null left
Add to Cart
 
Description

Description

May function as a transcription factor with dual roles in transcription initiation and termination. Binds to two sites, ME1a1 and ME1a2, within the MYC promoter having greater affinity for the former. Also binds to multiple G/C-rich sites within the promoter of the Sp1 family of transcription factors.
Specifications

Specifications

Maz
1 mg/mL
Monoclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
MAZ; MAZI; MYC associated zinc finger protein; Myc-associated zinc finger protein; MYC-associated zinc finger protein (purine-binding transcription factor); PUR1; Pur-1; Purine-binding transcription factor; SAF-1; SAF-2; SAF-3; serum amyloid A activating factor 1; serum amyloid A activating factor 2; Serum amyloid A-activating factor-1; Transcription factor Zif87; ZF87; Zif87; Zinc finger protein 801; zinc-finger protein, 87 kilodaltons; ZNF801
MAZ
IgG
Protein G
Antibody
133
Western Blot
Unconjugated
Mouse
Human
P56270
4150
This MAZ antibody was raised against recombinant human MAZ protein.
Primary
-20°C, Avoid Freeze/Thaw Cycles
MAZ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.