Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAZ/SAF-1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572382
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2382 10 μL
50-157-2381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2382 Supplier Bethyl Laboratories, Inc Supplier No. IHC00601T
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Monkey, Chimpanzee and Northern w

May function as a transcription factor with dual roles in transcription initiation and termination. Binds to two sites, ME1a1 and ME1a2, within the MYC promoter having greater affinity for the former. Also binds to multiple G/C-rich sites within the promoter of the Sp1 family of transcription factors.

Specifications

Antigen MAZ/SAF-1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MAZ
Gene Accession No. P56270
Gene Alias PUR1, Pur-1, SAF-1, SAF-2, SAF-3, ZF87, ZNF801, Zif87, MAZI, myc-associated zinc finger protein, purine-binding transcription factor, serum amyloid A activating factor 1, serum amyloid A activating factor 2, transcription factor Zif87, zinc finger protein 801, zinc-finger protein, 87 kilodaltons
Gene Symbols MAZ
Host Species Rabbit
Immunogen Between 427 and 477
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4150
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.