Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD1 Antibody (100B272.1), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB339592 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339592 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339592 Supplier Novus Biologicals Supplier No. NB10056537JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MBD1 Monoclonal antibody specifically detects MBD1 in Human, Mouse samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ChIP assay (ChIP), KnockDown

Specifications

Antigen MBD1
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 100B272.1
Conjugate Janelia Fluor 646
Dilution Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP), Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription
Host Species Mouse
Immunogen Generated by immunizing mice with a synthetic peptide corresponding to amino acids 391-405 (SESEDGAGSPPPYRR) of human MBD1, GenBank no ref NP_056671.2
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 4152
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.