Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD1 Antibody (10B10), Novus Biologicals™
SDP

Catalog No. NB298682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298682 50 μL
NB298683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298682 Supplier Novus Biologicals Supplier No. NBP32614750UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MBD1 Monoclonal antibody specifically detects MBD1 in Human samples. It is validated for ELISA, Immunocytochemistry

Specifications

Antigen MBD1
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 10B10
Conjugate Unconjugated
Dilution ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription
Host Species Rabbit
Immunogen A synthesized peptide derived from Human MBD1 [UniProt Q9UIS9]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 4152
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.