Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBD1 Antibody (10B10), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | MBD1 |
---|---|
Clone | 10B10 |
Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
MBD1 Monoclonal antibody specifically detects MBD1 in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
MBD1 | |
ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription | |
A synthesized peptide derived from Human MBD1 [UniProt Q9UIS9] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
10B10 | |
ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Chromatin Research | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4152 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title