Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB170506 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170506 25 μg
NB170507 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170506 Supplier Novus Biologicals Supplier No. NBP31867425UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MBD1 Polyclonal antibody specifically detects MBD1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen MBD1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.6 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:1000
Formulation PBS
Gene Accession No. Q9UIS9
Gene Alias CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human MBD1 (Methyl-CpG-binding domain protein 1), using 3 KLH-conjugated synthetic peptides, two containing a sequence from the C-terminal part and one containing a sequence from the N-terminal part of the protein.
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 4152
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.