Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB325686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325686 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325686 Supplier Novus Biologicals Supplier No. NBP299402F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MBD2 Polyclonal antibody specifically detects MBD2 in Human samples. It is validated for Western Blot

Specifications

Antigen MBD2
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias demethylase, DKFZp586O0821, DMTase, methyl-CpG binding domain protein 2, methyl-CpG-binding domain protein 2, Methyl-CpG-binding protein MBD2, NY-CO-41
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MBD2.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Chromatin Research, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8932
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.