Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD3 Antibody (PCRP-MBD3-1C4), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB380007 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380007 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380007 Supplier Novus Biologicals Supplier No. NBP308535G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MBD3 Monoclonal antibody specifically detects MBD3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen MBD3
Applications Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-MBD3-1C4
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias methyl-CpG binding domain protein 3, methyl-CpG-binding domain protein 3, Methyl-CpG-binding protein MBD3
Host Species Mouse
Immunogen Recombinant full-length human MBD3 protein (Uniprot: O95983)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 53615
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.