Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD3, Mouse anti-Human,Mouse, Clone: 106B691, Novus Biologicals™
SDP

Catalog No. NB013916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013916 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013916 Supplier Novus Biologicals Supplier No. NBP280843

Mouse Monoclonal Antibody

MBD3 Monoclonal specifically detects MBD3 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen MBD3
Applications ChIP Assay, CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 106B691
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 2 μg/mL, Chromatin Immunoprecipitation, Flow Cytometry : 1 ug/ million cells, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias methyl-CpG binding domain protein 3, methyl-CpG-binding domain protein 3, Methyl-CpG-binding protein MBD3
Gene Symbols MBD3
Host Species Mouse
Immunogen Between amino acids 200-250 of human MBD3 protein were used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 53615
Test Specificity Mouse reactivity reported in scientific literature (PMID: 17546630).
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.