Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224393 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224393 0.1 mg
NBP224393SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224393 Supplier Novus Biologicals Supplier No. NBP224393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

MBD4 Polyclonal specifically detects MBD4 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation (ChIP).

Specifications

Antigen MBD4
Applications Western Blot, ChIP Assay
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 15689352), Chromatin Immunoprecipitation (ChIP)
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias EC 3.2.2.-, G/T mismatch glycosylase, G/U mismatch glycosylase, MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics, methyl-CpG binding domain protein 4, methyl-CpG-binding domain protein 43,N(4)-ethenocytosine glycosylase, Methyl-CpG-binding endonuclease 1, Methyl-CpG-binding protein MBD4, Mismatch-specific DNA N-glycosylase, putative methyl-CpG binding protein
Gene Symbols MBD4
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 268-300 and 337-374 of human MBD4.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Base Excision Repair, Chromatin Research, DNA Repair, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 8930
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.