Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167010 20 μg
NB167009 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167010 Supplier Novus Biologicals Supplier No. NBP31595720UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MBD6 Polyclonal antibody specifically detects MBD6 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen MBD6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias methyl-CpG binding domain protein 6, methyl-CpG-binding domain protein 6, methyl-CpG-binding protein MBD6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 880-980 of human MBD6 (NP_443129.3).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114785
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.