Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBNL3 Antibody (PCRP-MBNL3-1D11), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB379237 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379237 Supplier Novus Biologicals Supplier No. NBP308663AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MBNL3 Monoclonal antibody specifically detects MBNL3 in Human samples. It is validated for Flow Cytometry, Protein Array

Specifications

Antigen MBNL3
Applications Flow Cytometry, Protein Array
Classification Monoclonal
Clone PCRP-MBNL3-1D11
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CHCRMBLX39FLJ11316, Cys3His CCG1-required protein, FLJ97142, MBXLMBLX, muscleblind-like 3 (Drosophila), muscleblind-like protein 3, Muscleblind-like X-linked protein, Protein HCHCR
Host Species Mouse
Immunogen Recombinant full-length human MBNL3 protein (Uniprot: Q9NUK0)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55796
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.