Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP Antibody (7G7), Novus Biologicals™
SDP

Catalog No. NBP105203 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105203 0.1 mL
NB018356 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP105203 Supplier Novus Biologicals Supplier No. NBP105203
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

MBP Monoclonal specifically detects MBP in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen MBP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 7G7
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation, Immunohistochemistry-Frozen
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Alias MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
Gene Symbols MBP
Host Species Mouse
Immunogen Purified myelin basic protein isolated from bovine brain, epitope is in peptide TPPPSQGKG, amino acids 125-133 of the human 21.5kDa sequence.
Molecular Weight of Antigen 19 kDa
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4155
Test Specificity The MBP Antibody (7G7) antibody binds all four of the CNS MBP isoforms, so that the epitope for the antibody is located in the core shared by all four gene products.
Target Species Human, Mouse, Rat, Pig, Bovine, Equine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.