Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBP Antibody (7G7), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$206.50 - $499.50
Specifications
Antigen | MBP |
---|---|
Clone | 7G7 |
Concentration | 1 mg/ml |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
MBP Monoclonal specifically detects MBP in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
MBP | |
1 mg/ml | |
Unconjugated | |
Mouse | |
Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
MBP | |
IgG1 | |
Affinity purified | |
The MBP Antibody (7G7) antibody binds all four of the CNS MBP isoforms, so that the epitope for the antibody is located in the core shared by all four gene products. |
7G7 | |
Monoclonal | |
Ascites | |
RUO | |
Pig, Equine, Human, Mouse, Rat, Bovine | |
4155 | |
Purified myelin basic protein isolated from bovine brain, epitope is in peptide TPPPSQGKG, amino acids 125-133 of the human 21.5kDa sequence. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
19 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title