Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB209314 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB209314 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB209314 Supplier Novus Biologicals Supplier No. NB11079873AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MBP Polyclonal antibody specifically detects MBP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry - Free Floating

Specifications

Antigen MBP
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
Host Species Rabbit
Immunogen Reacts with residues 49-62 of Isoforms 5 and 6 of the human protein. The immunogen corresponds to Isoform 1 between amino acids 182-195 of the 33 kDa isoform. The antibody also detects mouse and rat MBP protein. The antibody primarily detects a 22 kDa band.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4155
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.