Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP Antibody (MBP/4277R), FITC, Novus Biologicals™
SDP

Catalog No. NB380906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380906 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380906 Supplier Novus Biologicals Supplier No. NBP308388F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MBP Monoclonal antibody specifically detects MBP in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen MBP
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone MBP/4277R
Conjugate FITC
Formulation PBS
Gene Alias MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
Host Species Rabbit
Immunogen Recombinant fragment (around aa 150-250) of human MBP (exact sequence is proprietary) (Uniprot: P02686)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4155
Target Species Human, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.