Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP Antibody (MBP14) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB393083 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB393083 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB393083 Supplier Novus Biologicals Supplier No. NBP2619380.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MBP Monoclonal specifically detects MBP in Human, Bovine, Chicken, Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MBP
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MBP14
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:2000, Immunohistochemistry 1:100,000, Immunocytochemistry/Immunofluorescence 1:50
Formulation PBS with 0.02% Sodium Azide
Gene Alias MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
Gene Symbols MBP
Host Species Rat
Immunogen Reacts with human MBP, recognizing epitopes in the 36-50 amino acid region (GILDSIGRFFGGDRG)
Purification Method Protein G purified
Quantity 0.025 mg
Research Discipline Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4155
Target Species Bovine, Chicken, Equine, Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.