Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBP Antibody (MBP14) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$196.10
Specifications
Antigen | MBP |
---|---|
Clone | MBP14 |
Dilution | Western Blot 1:500, ELISA 1:2000, Immunohistochemistry 1:100,000, Immunocytochemistry/Immunofluorescence 1:50 |
Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Description
MBP Monoclonal specifically detects MBP in Human, Bovine, Chicken, Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MBP | |
Western Blot 1:500, ELISA 1:2000, Immunohistochemistry 1:100,000, Immunocytochemistry/Immunofluorescence 1:50 | |
Monoclonal | |
Purified | |
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
PBS with 0.02% Sodium Azide | |
4155 | |
Reacts with human MBP, recognizing epitopes in the 36-50 amino acid region (GILDSIGRFFGGDRG) | |
Primary |
MBP14 | |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rat | |
Bovine, Chicken, Equine, Human | |
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
MBP | |
IgG2b | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title