Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBTPS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094630 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094630 100 μL
NB094631 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094630 Supplier Novus Biologicals Supplier No. NBP305160100UL

Rabbit Polyclonal Antibody

MBTPS1 Polyclonal specifically detects MBTPS1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MBTPS1
Applications Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EC 3.4.21.112, Endopeptidase S1P, KIAA0091MGC138712, membrane-bound transcription factor peptidase, site 1, membrane-bound transcription factor protease, site 1, PCSK8, S1PMGC138711, site-1 protease, SKI1, SKI-1membrane-bound transcription factor site-1 protease, subtilisin/kexin isozyme-1, Subtilisin/kexin-isozyme 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 803-1052 of human MBTPS1 (NP_003782.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8720
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.