Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBTPS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | MBTPS1 |
---|---|
Applications | Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MBTPS1 Polyclonal specifically detects MBTPS1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MBTPS1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
8720 | |
IgG | |
Affinity purified |
Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
EC 3.4.21.112, Endopeptidase S1P, KIAA0091MGC138712, membrane-bound transcription factor peptidase, site 1, membrane-bound transcription factor protease, site 1, PCSK8, S1PMGC138711, site-1 protease, SKI1, SKI-1membrane-bound transcription factor site-1 protease, subtilisin/kexin isozyme-1, Subtilisin/kexin-isozyme 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 803-1052 of human MBTPS1 (NP_003782.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title