Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCAM/CD146 Antibody (012), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB099141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB099141 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB099141 Supplier Novus Biologicals Supplier No. NBP289202AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MCAM/CD146
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 12
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human MCAM/CD146 extracellular domain (NP_006491.2; Met 1-Gly 559) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4162
Test Specificity No cross-reactivity in ELISA with: Human CD66a Human CD166 Human CD106 Human BCAM Human CD226
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.