Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCAM/CD146 Antibody (733216), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB311165 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311165 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311165 Supplier Novus Biologicals Supplier No. FAB7718PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MCAM/CD146 Monoclonal antibody specifically detects MCAM/CD146 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen MCAM/CD146
Applications Flow Cytometry
Classification Monoclonal
Clone 733216
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MCAM/CD146, Met1-Val563, Accession # Q8R2Y2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4162
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.