Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCAM/CD146 Rabbit anti-Mouse, Clone: 112, Novus Biologicals™
SDP

Catalog No. NB025184 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB025184 50 μg
NB025183 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB025184 Supplier Novus Biologicals Supplier No. NBP29071850UL

Rabbit Monoclonal Antibody

MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Mouse samples. It is validated for ELISA.

Specifications

Antigen MCAM/CD146
Applications ELISA
Classification Monoclonal
Clone 112
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin
Host Species Rabbit
Immunogen Recombinant Mouse MCAM/CD146 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4162
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.