Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCF2L2 Antibody, Novus Biologicals™
SDP

Catalog No. NB301478 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301478 Supplier Novus Biologicals Supplier No. NBP324974
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MCF2L2 Polyclonal antibody specifically detects MCF2L2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MCF2L2
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias ARHGEF22, dbs-related Rho family guanine nucleotide exchange factor, MCF.2 cell line derived transforming sequence-like 2, MCF2-transforming sequence-like protein 2
Host Species Rabbit
Immunogen This antibody has been engineered to specifically recognize the recombinant protein MCF2L2 using the following amino acid sequence: MLSTEDLLMSHTRQRDKLQDELKLLGKQGTTLLSCIQEPATKCPNSKLNLNQLENVTTMERLLVQLDETEKAFSHFWSEHHLKLNQCLQLQHFEHDF
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23101
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.