Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCG10 Antibody, Novus Biologicals™
SDP

Catalog No. NB035411 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB035411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB035411 Supplier Novus Biologicals Supplier No. NBP287789
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MCG10 Polyclonal specifically detects MCG10 in Human samples. It is validated for Western Blot.

Specifications

Antigen MCG10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias alpha-CP4, alphaCP-4 protein, CBP, LIP4, LYST-interacting protein, MCG10, poly(rC) binding protein 4, poly(rC)-binding protein 4, RNA binding protein MCG10
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human MCG10
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 57060
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.