Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCG10 Antibody, Novus Biologicals™
SDP

Catalog No. NB10068152 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068152 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068152 Supplier Novus Biologicals Supplier No. NB10068152
Add to Cart
Add to Cart

Goat Polyclonal Antibody

MCG10 Polyclonal specifically detects MCG10 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen MCG10
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_065151.1
Gene Alias alpha-CP4, alphaCP-4 protein, CBP, LIP4, LYST-interacting protein, MCG10, poly(rC) binding protein 4, poly(rC)-binding protein 4, RNA binding protein MCG10
Gene Symbols PCBP4
Host Species Goat
Immunogen Peptide with sequence C-NGSKKAERQKFSPY corresponding to C-Terminus according to NP_127501.1, NP_127502.1, NP_127503.1 .
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 57060
Test Specificity This antibody is expected to recognize all reported isoforms (NP_065151.1, NP_127501.1, NP_127502.1 and NP_127503.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.