Learn More
Description
Specifications
Specifications
| Antigen | mCherry |
| Applications | Western Blot, Immunohistochemistry, Immunoassay |
| Classification | Monoclonal |
| Clone | 1C51 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, Immunohistochemistry, Knockout Validated |
| Gene Alias | DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein |
| Host Species | Mouse |
| Immunogen | This mCherry Antibody (1C51) was developed against recombinant full-length mCherry purified from E. coli. |
| Molecular Weight of Antigen | 27 kDa |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
