Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mCherry Antibody (2783C), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB305362 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305362 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305362 Supplier Novus Biologicals Supplier No. FAB11041J
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

mCherry Monoclonal antibody specifically detects mCherry in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunocytochemistry

Specifications

Antigen mCherry
Applications Western Blot, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 2783C
Conjugate Janelia Fluor 646
Dilution Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
Host Species Rabbit
Immunogen Human mCherry synthetic peptide
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Fusion Proteins
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.