Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCL-1 Monoclonal Antibody (Ab22), eBioscience™
GREENER_CHOICE

Catalog No. 5013517
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-135-17 100 μg
14-670-137 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-135-17 Supplier Invitrogen™ Supplier No. 14670182
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This Ab22 monoclonal antibody reacts with the human myeloid cell leukemia-1 (Mcl-1) protein. This 40 kDa Bcl-2 family member is expressed in B and T cells, macrophages, neutrophils, and hematopoietic stem cells. Mcl-1 is also expressed in non-immune cells such as fibroblasts, epithelial cells, neuroendocrine cells, chondrocytes, and hepatocytes. Reportedly induced by growth factors and cytokines in normal cells, Mcl-1 expression of Mcl-1 can be dysregulated in various leukemias and cancer. Mcl-1 mediates cell survival and proliferation by sequestering the pro-apoptotic Bcl-2 proteins Bak, Bax, and Bim. This protein is also involved in germinal center and memory B cell formation, as well as in myeloid and early lymphoid differentiation. Finally, Mcl-1 activity and dimerization with Bcl-2 family members is dependent on serine and threonine phosphorylation. Applications Reported: This Ab22 antibody has been reported for use in immunoblotting (WB) and immunocytochemistry (ICC). Applications Tested: This Ab22 antibody has been tested by immunocytochemistry on fixed and permeabilized MCF7 cells at less than or equal to 5 μg/mL. This Ab22 has also been tested by western blot of reduced Jurkat cell lysate and can be used at 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm...

MCL1 (Myeloid cell leukemia-1) belongs to the Bcl-2 family and is involved in programing, differentiation and concomitant maintenance of cell viability, but not of proliferation. Isoform 1 of MCL1 inhibits apoptosis while isoform 2 promotes it. The carboxy terminal of MCL1 and bcl-2 share significant sequence homology. Expression of MCL1 is increased upon exposure of ML-1 cells to various types of DNA damaging agents (e.g. ionizing radiation, ultraviolet radiation, and alkylating drugs) along with increases in GADD45 and Bax and a decrease in bcl-2. Enhanced expression of MCL1, prominently associated with mitochondria, complements the continued expression of bcl-2 in ML-1 cells undergoing differentiation. Like bcl-2, MCL1 has the capacity to promote cell viability under conditions that otherwise cause apoptosis. While the mechanism by which MCL1 inhibits apoptosis is not known, it is thought that it may heterodimerize and neutralize pro-apoptotic members of the Bcl-2 family such as Bim or Bak. MCL1 was originally identified in differentiating myeloid cells, but has since been shown to be expressed in multiple cell types. MCL1 is essential for embryogenesis and for the development and maintenance of B and T lymphocytes in animals. MCL1 exists as at least two distinct isoforms designated MCL1L and MCL1S. In marked contrast to the larger isoform of MCL1, overexpression of MCL1S promotes cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCL-1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone Ab22
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene MCL1
Gene Accession No. Q07820
Gene Alias AW556805; BCL2 family apoptosis regulator; BCL2L3; bcl2-L-3; Bcl-2-like protein 3; bcl-2-related protein EAT/mcl1; EAT; Induced myeloid leukemia cell differentiation protein Mcl-1; induced myeloid leukemia cell differentiation protein Mcl-1 homolog; Mcl1; Mcl-1; mcl1/EAT; MCL1-ES; MCL1L; MCL1S; MCL-1S; MGC104264; MGC1839; myeloid cell leukemia 1; myeloid cell leukemia ES; myeloid cell leukemia sequence 1; myeloid cell leukemia sequence 1 (BCL2-related); TM
Gene Symbols MCL1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4170
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.